Diethylamine hydrochloride 99%

Warning

Synonyms: Diethylammonium chloride

AC15589-0010 AC15589-5000 AC15589-0051 AC15589-1000
200018-794EA 57.7 USD
200018-794 200012-068 103510-636 200043-240
Diethylamine hydrochloride 99%
Diethylamine hydrochloride
Formula: C₄H₁₂ClN
MW: 109.6 g/mol
Boiling Pt: 320…330 °C (760 mmHg)
Melting Pt: 227…230 °C
Storage Temperature: Ambient
MDL Number: MFCD00012499
CAS Number: 660-68-4
EINECS: 211-541-9
Merck Index: 13,03138

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR