3-Aminobenzoic acid 99+%

Warning

AC15585-0250 AC15585-1000 AC15585-5000
200035-246EA 152 USD
200035-246 200016-940 200009-770
3-Aminobenzoic acid 99+%
3-Aminobenzoic acid
Formula: C₇H₇NO₂
MW: 137.14 g/mol
Boiling Pt: 308 °C (1013 hPa)
Melting Pt: >172 °C
Flash Pt: 150 °C
Storage Temperature: Ambient
MDL Number: MFCD00007795
CAS Number: 99-05-8
EINECS: 202-724-4
Merck Index: 13,00420

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR