2-Methylbenzylamine 96%

Danger

Synonyms: 2-(Aminomethyl)tolueneo-Tolylmethanamine

AC15563-0050 AC15563-0250
103531-438EA 261 USD
103531-438 200016-684
2-Methylbenzylamine 96%
2-Methylbenzylamine
Formula: C₈H₁₁N
MW: 121.18 g/mol
Boiling Pt: 196…197 °C (760 mmHg)
Melting Pt: < 0 °C
Density: 0.97 g/cm³
Flash Pt: 83 °C
Storage Temperature: Ambient
MDL Number: MFCD00008112
CAS Number: 89-93-0
EINECS: 201-952-1
UN: 2735
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR