1,2-Bis(diethylamino)ethane 99+%

Danger

Synonyms: N,N,N',N'-TetraethylethylenediamineN1,N1,N2,N2-Tetraethylethane-1,2-diamine

AC15559-0100 AC15559-5000 AC15559-1000
103531-436EA 998 USD
103531-436 200032-044 200013-490
1,2-Bis(diethylamino)ethane 99+%
1,2-Bis(diethylamino)ethane
Formula: C₁₀H₂₄N₂
MW: 172.31 g/mol
Boiling Pt: 189…192 °C (760 mmHg)
Density: 0.808 g/cm³
Flash Pt: 58 °C
Storage Temperature: Ambient
MDL Number: MFCD00009055
CAS Number: 150-77-6
EINECS: 205-770-3
UN: 2734
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR