3,4-Xylidine 99.5%

Danger

Synonyms: 3,4-Dimethylaniline4-Amino-o-xylene

AC15528-0050 AC15528-1000 AC15528-5000
200009-678EA 34.8 USD
200009-678 200047-260 200029-092
3,4-Xylidine 99.5%
3,4-Xylidine
Formula: C₈H₁₁N
MW: 121.18 g/mol
Boiling Pt: 226 °C (1013 hPa)
Melting Pt: 49…51 °C
Flash Pt: 98 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007810
CAS Number: 95-64-7
EINECS: 202-437-4
UN: 3452
ADR: 6.1,II
Merck Index: 12,10217

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR