1,3-Dimethoxybenzene 99%

Synonyms: Resorcinol dimethyl ether

AC15523-1000 AC15523-5000
200021-650EA 106 USD
200021-650 200008-476
1,3-Dimethoxybenzene 99%
1,3-Dimethoxybenzene
Formula: C₈H₁₀O₂
MW: 138.17 g/mol
Boiling Pt: 85…87 °C (7 mmHg)
Melting Pt: –52 °C
Density: 1.05 g/cm³
Flash Pt: 87 °C
Storage Temperature: Ambient
MDL Number: MFCD00008384
CAS Number: 151-10-0
EINECS: 205-783-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR