2,3-Diaminopyridine 98%

Danger

Synonyms: Pyridine-2,3-diyldiamine

AC15502-0250 AC15502-0050 AC15502-1000
103531-404EA 865.85 USD
103531-404 200004-866 103531-400
2,3-Diaminopyridine 98%
2,3-Diaminopyridine
Formula: C₅H₇N₃
MW: 109.13 g/mol
Boiling Pt: 148…150 °C (5 mmHg)
Melting Pt: 114…116 °C
Flash Pt: 205 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006319
CAS Number: 452-58-4
EINECS: 207-200-9
UN: 2811
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR