1,4-Cyclohexanedione 98%

AC15483-0100 AC15483-0500 AC15483-2500
200015-586EA 32.2 USD
200015-586 200008-496 103510-616
1,4-Cyclohexanedione 98%
1,4-Cyclohexanedione
Formula: C₆H₈O₂
MW: 112.13 g/mol
Boiling Pt: 112 °C (26hPa)
Melting Pt: 76…80 °C
Flash Pt: 132 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00001606
CAS Number: 637-88-7
EINECS: 211-306-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR