Citraconic anhydride 98%

Danger

Synonyms: 3-Methylfuran-2,5-dione

AC15479-2500 AC15479-1000 AC15479-0250 AC15479-0010
103531-390EA 432 USD
103531-390 200018-544 103531-392 200011-706
Citraconic anhydride 98%
Citraconic anhydride
Formula: C₅H₄O₃
MW: 112.08 g/mol
Boiling Pt: 213 °C (760 mmHg)
Melting Pt: 6…7 °C
Density: 1.245 g/cm³
Flash Pt: 101 °C
Storage Temperature: Ambient
MDL Number: MFCD00005522
CAS Number: 616-02-4
EINECS: 210-459-0
UN: 2922
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR