4-Chlorothiophenol 98%

Danger

Synonyms: 1-Chloro-4-mercaptobenzene4-Chlorobenzenethiolp-Chlorothiophenol4-Chlorobenzene-1-thiol

AC15476-1000 AC15476-5000 AC15476-0050
200017-488EA 26.8 USD
200017-488 200041-962 200010-360
4-Chlorothiophenol 98%
4-Chlorothiophenol
Formula: C₆H₅ClS
MW: 144.62 g/mol
Boiling Pt: 205 °C (1013 hPa)
Melting Pt: 49…53 °C
Flash Pt: 109 °C
Storage Temperature: Ambient
MDL Number: MFCD00004847
CAS Number: 106-54-7
EINECS: 203-408-9
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR