Chloro(diphenyl)methane 98%

Danger

Synonyms: Benzhydryl chloride

AC15466-0500 AC15466-2500 AC15466-0050
103531-386EA 75.8 USD
103531-386 200024-732 200042-880
Chloro(diphenyl)methane 98%
Chloro(diphenyl)methane
Formula: (C₆H₅)₂CHCl
MW: 202.68 g/mol
Boiling Pt: 270 °C (760 mmHg)
Melting Pt: 15…17 °C
Density: 1.14 g/cm³
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00000855
CAS Number: 90-99-3
EINECS: 202-031-7
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR