2-Chlorobenzaldehyde 99%

Danger

AC15461-1000 AC15461-0010 AC15461-0050 AC15461-5000 AC15461-2500
200005-024EA 24.6 USD
200005-024 200028-640 200016-486 200040-940 200016-484
2-Chlorobenzaldehyde 99%
2-Chlorobenzaldehyde
Formula: C₇H₅ClO
MW: 140.57 g/mol
Boiling Pt: 211.9 °C (1013 hPa)
Melting Pt: 10…11.5 °C
Density: 1.25 g/cm³
Flash Pt: 87 °C
Storage Temperature: Ambient
MDL Number: MFCD00003304
CAS Number: 89-98-5
EINECS: 201-956-3
UN: 3265
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR