Glycolic acid 99%

Danger

Synonyms: 2-Hydroxy acid2-Hydroxyacetic acid

AC15451-0250 AC15451-1000 AC15451-5000 AC15451-0025
200001-278EA 48.5 USD
200001-278 103525-840 200001-280 200001-282
Glycolic acid 99%
Glycolic acid
Formula: C₂H₄O₃
MW: 76.05 g/mol
Boiling Pt: 100 °C (760 mmHg)
Melting Pt: 72…80 °C
Flash Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00004312
CAS Number: 79-14-1
EINECS: 201-180-5
UN: 3261
ADR: 8,II
Merck Index: 13,04511

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR