(4-Bromophenyl)acetonitrile 99%

Danger

Synonyms: 4-Bromobenzyl cyanide4-Bromophenylacetonitrile

AC15448-0250 AC15448-1000
200029-660EA 34.6 USD
200029-660 200010-290
(4-Bromophenyl)acetonitrile 99%
(4-Bromophenyl)acetonitrile
Formula: C₈H₆BrN
MW: 196.05 g/mol
Boiling Pt: 286 °C (1013 hPa)
Melting Pt: 47…49 °C
Flash Pt: 152 °C
MDL Number: MFCD00001916
CAS Number: 16532-79-9
EINECS: 240-602-2
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR