Butyrophenone 99%

Synonyms: 1-Phenyl-1-butanone1-Phenylbutan-1-onePhenyl propyl ketone

AC15441-5000 AC15441-1000
103510-608EA 277 USD
103510-608 200001-276
Butyrophenone 99%
Butyrophenone
Formula: C₆H₅C(O)(CH₂)₂CH₃
MW: 148.2 g/mol
Boiling Pt: 228…230 °C (760 mmHg)
Melting Pt: 11…13 °C
Density: 0.988 g/cm³
Flash Pt: 88 °C
Storage Temperature: Ambient
MDL Number: MFCD00009397
CAS Number: 495-40-9
EINECS: 207-799-7
https://us.vwr.com/stibo/web/std.lang.all/22/17/762217.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR