3-tert-Butylphenol 99+%

Danger

Synonyms: m-tert-Butylphenol

AC15438-1000 AC15438-0250 AC15438-0010
200010-078EA 116 USD
200010-078 103531-384 200023-458
3-tert-Butylphenol 99+%
3-tert-Butylphenol
Formula: C₁₀H₁₄O
MW: 150.22 g/mol
Boiling Pt: 125…130 °C (20 mmHg)
Melting Pt: 40…43 °C
Flash Pt: 108 °C
Storage Temperature: Ambient
MDL Number: MFCD00002300
CAS Number: 585-34-2
EINECS: 209-553-4
UN: 2923
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR