1-Bromo-3-chlorobenzene 99%
|
Warning
|
Synonyms:
3-Bromochlorobenzene
AC15420-1000
AC15420-0250
200023-198EA
34.5
USD
200023-198
200035-280
1-Bromo-3-chlorobenzene 99%
1-Bromo-3-chlorobenzene
Formula:
C₆H₄BrCl MW: 191.45 g/mol Boiling Pt: 196 °C (1013 hPa) Melting Pt: –21.5 °C Density: 1.63 g/cm³ Flash Pt: 80 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000568 CAS Number: 108-37-2 EINECS: 203-575-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...