4-(Benzyloxy)benzaldehyde 98%

AC15411-0250
200010-216EA 41.3 USD
200010-216
4-(Benzyloxy)benzaldehyde 98%
4-(Benzyloxy)benzaldehyde
Formula: C₁₄H₁₂O₂
MW: 212.25 g/mol
Boiling Pt: 197…199 °C (11 mmHg)
Melting Pt: 70…74 °C
Storage Temperature: Ambient
MDL Number: MFCD00003387
CAS Number: 4397-53-9
EINECS: 224-527-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR