Anisole 99%, pure

Warning

Synonyms: MethoxybenzeneMethyl phenyl ether

AC15392-0050 AC15392-0010 AC15392-2500 AC15392-0025
200011-246EA 41 USD
200011-246 200001-262 103525-828 200001-260
Anisole 99%, pure
Anisole
Formula: C₇H₈O
MW: 108.14 g/mol
Boiling Pt: 154 °C (760 mmHg)
Melting Pt: –37 °C
Density: 0.99 g/cm³
Flash Pt: 45 °C
Storage Temperature: Ambient
MDL Number: MFCD00008354
CAS Number: 100-66-3
EINECS: 202-876-1
UN: 2222
ADR: 3,III
Merck Index: 13,00672
https://us.vwr.com/stibo/web/std.lang.all/96/93/759693.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR