4-Aminohippuric acid 99%

Synonyms: p-Aminohippuric acid2-(4-Aminobenzamido)acetic acid

AC15380-0100 AC15380-1000 AC15380-0250 AC15380-5000
200017-366EA 71.3 USD
200017-366 103510-592 103510-594 200017-368
4-Aminohippuric acid 99%
4-Aminohippuric acid
Formula: H₂NC₆H₄C(O)NHCH₂COOH
MW: 194.19 g/mol
Melting Pt: 198…202 °C
Storage Temperature: Ambient
MDL Number: MFCD00007890
CAS Number: 61-78-9
EINECS: 200-518-9
Merck Index: 13,00441

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR