2-Amino-3-hydroxypyridin 98%

Danger

Synonyms: 2-Amino-3-hydroxypyridine2-Aminopyridin-3-ol

AC15357-1000 AC15357-0250
200022-388EA 171 USD
200022-388 200046-642
2-Amino-3-hydroxypyridin 98%
2-Amino-3-hydroxypyridin
Formula: C₅H₆N₂O
MW: 110.12 g/mol
Boiling Pt: 250 °C (1013 hPa)
Melting Pt: 170…176 °C
Storage Temperature: Ambient
MDL Number: MFCD00006317
CAS Number: 16867-03-1
EINECS: 240-886-8
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR