Chroman-4-one 96%

Synonyms: 4-Chromanone

AC15346-0100 AC15346-0500
200010-362EA 253.81 USD
200010-362 200029-768
Chroman-4-one 96%
Chroman-4-one
Formula: C₉H₈O₂
MW: 148.16 g/mol
Boiling Pt: 127…128 °C (13 mmHg)
Melting Pt: 34…40 °C
Flash Pt: 110 °C
MDL Number: MFCD00006840
CAS Number: 491-37-2
EINECS: 207-736-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR