3,5-Dibromosalicylaldehyde 98%

Warning

Synonyms: 3,5-Dibromo-2-hydroxybenzaldehyde

AC15340-0010 AC15340-1000 AC15340-0250 AC15340-5000
200041-394EA 302 USD
200041-394 103465-122 200041-396 103465-124
3,5-Dibromosalicylaldehyde 98%
3,5-Dibromosalicylaldehyde
Formula: C₇H₄Br₂O₂
MW: 279.92 g/mol
Melting Pt: 80…84 °C
Storage Temperature: Ambient
MDL Number: MFCD00003318
CAS Number: 90-59-5
EINECS: 202-003-4
Merck Index: 13,03052

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR