2,6-Dichlorophenolindophenol sodium salt hydrate 98+%

Synonyms: Sodium 4-(3,5-dichloro-4-oxocyclohexa-2,5-dienylideneamino)phenoxide hydrateDPIP2,6-Dichlorophenolindophenol sodium salt xhydrateDCIPSodium 2,6-dichloroindophenolate hydrate2,6-Dichloroindophenol, sodium salt hydrate2,6-Dichloroindophenol sodium hydrate2,6-Dichloro-N-(4-hydroxyphenyl)-1,4-benzoquinoneimine sodium salt hydrateTillmans' reagent hydrate

AC15287-0100 AC15287-0010 AC15287-1000
200009-062EA 26 USD
200009-062 103466-902 200004-906
2,6-Dichlorophenolindophenol sodium salt hydrate 98+%
2,6-Dichlorophenolindophenol sodium salt hydrate
Formula: C₁₂H₆Cl₂NNaO₂·xH₂O
MW: 290.08 g/mol (anhy)
Storage Temperature: Ambient
MDL Number: MFCD00150014
CAS Number: 1266615-56-8
EINECS: 210-640-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR