2,4,6-Triisopropylbenzenesulfonyl chloride 97%

Danger

Synonyms: 2,4,6-Triisopropylbenzene-1-sulfonyl chloride2,4,6-Triisopropylbenzene-1-sulphonyl chloride

AC15279-1000 AC15279-0250
200008-946EA 82.2 USD
200008-946 200022-154
2,4,6-Triisopropylbenzenesulfonyl chloride 97%
2,4,6-Triisopropylbenzenesulphonyl chloride
Formula: C₁₅H₂₃ClO₂S
MW: 302.87 g/mol
Melting Pt: 93…97 °C
Storage Temperature: Ambient
MDL Number: MFCD00007433
CAS Number: 6553-96-4
EINECS: 229-479-6
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR