p-Anisic acid 98%

Synonyms: 4-Methoxybenzoic acid4-Anisic acid

AC15237-2500 AC15237-0051 AC15237-1000 AC15237-0010
103466-266EA 687 USD
103466-266 200050-380 200020-300 200007-266
p-Anisic acid 98%
p-Anisic acid
Formula: C₈H₈O₃
MW: 152.15 g/mol
Boiling Pt: 275 °C (760 mmHg)
Melting Pt: 181…186 °C
Flash Pt: 185 °C
Storage Temperature: Ambient
MDL Number: MFCD00002542
CAS Number: 100-09-4
EINECS: 202-818-5
Merck Index: 13,00669

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR