2-Methyl-1,3-cyclopentanedione 98%

Synonyms: 2-Methylcyclopentane-1,3-dione

AC15232-0250 AC15232-1000
103464-754EA 347 USD
103464-754 200009-478
2-Methyl-1,3-cyclopentanedione 98%
2-Methyl-1,3-cyclopentanedione
Formula: C₆H₈O₂
MW: 112.13 g/mol
Melting Pt: 212…216 °C
Storage Temperature: Ambient
MDL Number: MFCD00001406
CAS Number: 765-69-5
EINECS: 212-153-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR