1-Benzylimidazole 99%

Synonyms: 1-Benzyl-1H-imidazole

AC15225-0050 AC15225-0250
200021-750EA 34.9 USD
200021-750 200008-578
1-Benzylimidazole 99%
1-Benzylimidazole
Formula: C₁₀H₁₀N₂
MW: 158.2 g/mol
Boiling Pt: 310 °C (760 mmHg)
Melting Pt: 68…71 °C
Storage Temperature: Ambient
MDL Number: MFCD00005296
CAS Number: 4238-71-5
EINECS: 224-200-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR