2-Chloro-6-methylpyridine 98%

Warning

Synonyms: 6-Chloro-2-picoline

AC15223-0100 AC15223-0010
103465-766EA 18 USD
103465-766 200024-158
2-Chloro-6-methylpyridine 98%
2-Chloro-6-methylpyridine
Formula: C₆H₆ClN
MW: 127.57 g/mol
Boiling Pt: 64…68 °C (10 mmHg)
Density: 1.16 g/cm³
Flash Pt: 73 °C
Storage Temperature: Ambient
MDL Number: MFCD00006245
CAS Number: 18368-63-3
EINECS: 242-241-6
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR