2-Chloro-3-hydroxypyridine 98%

Warning

Synonyms: 2-Chloro-3-pyridinol

AC15218-0250 AC15218-1000
103464-460EA 315.1 USD
103464-460 200016-450
2-Chloro-3-hydroxypyridine 98%
2-Chloro-3-hydroxypyridine
Formula: C₅H₄ClNO
MW: 129.55 g/mol
Melting Pt: 168…172 °C
Storage Temperature: Ambient
MDL Number: MFCD00006235
CAS Number: 6636-78-8
EINECS: 229-635-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR