2,2'-Dihydroxybiphenyl 99%

Warning

Synonyms: 2,2'-BiphenolBiphenyl-2,2'-diol2,2-Biphenol

AC15199-5000 AC15199-1000
200022-032EA 47 USD
200022-032 200034-132
2,2'-Dihydroxybiphenyl 99%
2,2'-Dihydroxybiphenyl
Formula: C₁₂H₁₀O₂
MW: 186.21 g/mol
Boiling Pt: 315 °C (1013 hPa)
Melting Pt: 108…111 °C
Density: 0.83 g/cm³
Flash Pt: 154 °C
MDL Number: MFCD00002210
CAS Number: 1806-29-7
EINECS: 217-303-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR