Hexanal 96% stabilized

Warning

Synonyms: Hexyl aldehydeAldehyde C6CaproaldehydeHexylaldehydeHexaldehyde

AC15185-0025 AC15185-0010 AC15185-5000
200012-674EA 121 USD
200012-674 103525-802 200037-602
Hexanal 96% stabilized
Hexanal
Formula: C₆H₁₂O
MW: 100.16 g/mol
Boiling Pt: 130…131 °C (760 mmHg)
Melting Pt: –56 °C
Density: 0.815
Flash Pt: 32 °C
Storage Temperature: Ambient
MDL Number: MFCD00007027
CAS Number: 66-25-1
EINECS: 200-624-5
UN: 1207
ADR: 3,III
Merck Index: 13,01766

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR