Fluoroacetone 97%

Danger

AC15178-0010 AC15178-0050
200055-620EA 64.8 USD
200055-620 103531-356
Fluoroacetone 97%
Fluoroacetone
Formula: C₃H₅FO
MW: 76.07 g/mol
Boiling Pt: 75 °C (760 mmHg)
Density: 1.054 g/cm³
Flash Pt: 7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000451
CAS Number: 430-51-3
EINECS: 207-064-0
UN: 1992
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR