Dodecylbenzene 97%

Synonyms: 1-Phenyldodecane

AC15102-0500 AC15102-0100
200034-018EA 42.5 USD
200034-018 200008-778
Dodecylbenzene 97%
Dodecylbenzene
Formula: C₁₈H₃₀
MW: 246.44 g/mol
Boiling Pt: 288…300 °C (760 mmHg)
Melting Pt: –7 °C
Density: 0.85 g/cm³
Flash Pt: 124 °C
Storage Temperature: Ambient
MDL Number: MFCD00008974
CAS Number: 123-01-3
EINECS: 204-591-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR