Octylbenzene 99%

Synonyms: 1-Phenyloctane

AC15099-0250
200008-782EA 82 USD
200008-782
Octylbenzene 99%
Octylbenzene
Formula: C₁₄H₂₂
MW: 190.33 g/mol
Boiling Pt: 264 °C (1013 hPa)
Melting Pt: –36 °C
Density: 0.85 g/cm³
Flash Pt: 107 °C
Storage Temperature: Ambient
MDL Number: MFCD00009564
CAS Number: 2189-60-8
EINECS: 218-582-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR