Tributyrin (glycerol tributyrate) ≥98%

Supplier: ACROS ORGANICS MS

Synonyms: Glycerol tributyrateGlyceryl tributyrate

AC15088-2500 AC15088-1000 AC15088-0010
200001-204EA 59.65 USD InStock
200001-204 200007-938 103510-534
Tributyrin (glycerol tributyrate) ≥98%
Tributyrin (Glycerol tributyrate)
Formula: C₁₅H₂₆O₆
MW: 302.37 g/mol
Boiling Pt: 305°C - 310°C (760mmHg)
Melting Pt: -70 °C
Density: 1.035 g/cm³ (20 °C)
Flash Pt: 173°C
Storage Temperature: Ambient
MDL Number: MFCD00009392
CAS Number: 60-01-5
EINECS: 200-451-5
Merck Index: 12,09750

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4340 to 1.4370 (20°C, 589 nm)
GC ≥97.5 %
Color scale ≤70 APHA
Identification Passes test
Specific gravity (20°C) 1.032 to 1.035

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR