Acetanilide 99+%

Warning

Synonyms: N-Phenylacetamide

AC15081-5000 AC15081-0051 AC15081-0050 AC15081-0010 AC15081-2500
200001-196EA 52.7 USD
200001-196 200001-198 103510-530 200010-902 200010-900
Acetanilide 99+%
Acetanilide
Formula: C₈H₉NO
MW: 135.17 g/mol
Boiling Pt: 304 °C (760 mmHg)
Melting Pt: 111…115 °C
Flash Pt: 173 °C
Storage Temperature: Ambient
MDL Number: MFCD00008674
CAS Number: 103-84-4
EINECS: 203-150-7
Merck Index: 13,00051

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR