4-Nitro-o-anisidine 99%

Warning

Synonyms: 2-Methoxy-4-nitroaniline2-Amino-5-nitroanisole

AC15064-5000 AC15064-1000 AC15064-0050
200028-830EA 34 USD
200028-830 200041-124 103463-566
4-Nitro-o-anisidine 99%
4-Nitro-o-anisidine
Formula: C₇H₈N₂O₃
MW: 168.15 g/mol
Melting Pt: 139…142 °C
Storage Temperature: Ambient
MDL Number: MFCD00007363
CAS Number: 97-52-9
EINECS: 202-588-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR