1-Octanol 99%, pure

Warning

Synonyms: n-Octanol

AC15063-0025 AC15063-0010 AC15063-2500
200001-178EA 83.3 USD
200001-178 103510-524 200008-752
1-Octanol 99%, pure
1-Octanol
Formula: H₃C(CH₂)₆CH₂OH
MW: 130.23 g/mol
Boiling Pt: 195 °C (760 mmHg)
Melting Pt: –16 °C
Density: 0.824 g/cm³
Flash Pt: 81 °C
Storage Temperature: Ambient
MDL Number: MFCD00002988
CAS Number: 111-87-5
EINECS: 203-917-6
Merck Index: 12,06849
https://us.vwr.com/stibo/web/std.lang.all/14/03/731403.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR