D-(+)-Galactose 99+%

Synonyms: (2R,3S,4S,5R)-2,3,4,5,6-Pentahydroxyhexanal(2R,3S,4S,5R)-Pentahydroxyhexanal

AC15061-5000 AC15061-1000 AC15061-0010
200001-174EA 68.29 USD
200001-174 200001-176 200001-172
D-(+)-Galactose 99+%
D(+)-Galactose
Formula: C₆H₁₂O₆
MW: 180.16 g/mol
Boiling Pt: 527 °C (1013 hPa)
Melting Pt: 165…168 °C
Storage Temperature: Ambient
MDL Number: MFCD00151230
CAS Number: 59-23-4
EINECS: 200-416-4
Merck Index: 13,04356
https://us.vwr.com/stibo/web/std.lang.all/21/21/2302121.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR