D-(+)-Mannose 99+%
Synonyms:
(2S,3S,4R,5R)-2,3,4,5,6-Pentahydroxyhexanal
AC15060-0010
AC15060-0250
AC15060-1000
200030-860EA
864
USD
200030-860
200001-168
200001-170
D-(+)-Mannose 99+%
D(+)-Mannose
Formula:
C₆H₁₂O₆ MW: 180.16 g/mol Melting Pt: 133 °C Storage Temperature: Ambient |
MDL Number:
MFCD00064122 CAS Number: 3458-28-4 EINECS: 222-392-4 Merck Index: 13,05772 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...