4-Ethylpyridine

Supplier: ACROS ORGANICS MS

Danger

AC15057-5000 AC15057-0250 AC15057-1000
200041-994EA 257.46 USD
200041-994 200041-992 200029-808
4-Ethylpyridine
4-Ethylpyridine
Formula: C₇H₉N
MW: 107.16 g/mol
Boiling Pt: 168,3 °C (1013 hPa)
Melting Pt: –91 °C
Density: 0.949 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006450
CAS Number: 536-75-4
EINECS: 208-646-7
UN: 1993
ADR: 3,III
Merck Index: 13,03880

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5009 to 1.5029 (20°C, 589 nm)
Water ≤0.5 % (K.F.)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR