Ethyl trans-cinnamate 98% (trans-isomers)

AC15053-5000 AC15053-0025 AC15053-1000
200001-164EA 110 USD
200001-164 200025-340 103510-516
Ethyl trans-cinnamate 98% (trans-isomers)
Ethyl trans-cinnamate
Formula: C₁₁H₁₂O₂
Boiling Pt: 271 °C (760 mmHg)
Melting Pt: 6.5…7.5 °C
Density: 1.046 g/cm³
Flash Pt: 135 °C
Storage Temperature: Ambient
MDL Number: MFCD00009189
CAS Number: 103-36-6
EINECS: 203-104-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR