Ethyl octanoate 99+%

Synonyms: Caprylic acid ethyl esterOctanoic acid ethyl esterEthyl caprylate

AC15051-1000 AC15051-0050 AC15051-5000
200043-474EA 16 USD
200043-474 200025-334 200031-276
Ethyl octanoate 99+%
Ethyl octanoate
Formula: C₁₀H₂₀O₂
MW: 172.27 g/mol
Boiling Pt: 206 °C (1013 hPa)
Melting Pt: –47 °C
Density: 0.868 g/cm³
Flash Pt: 75 °C
Storage Temperature: Ambient
MDL Number: MFCD00009552
CAS Number: 106-32-1
EINECS: 203-385-5
Merck Index: 13,03813

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR