α,4-Dibromotoluene 98%

Danger

Synonyms: 4-Bromobenzyl bromide1-Bromo-4-(bromomethyl)benzene

AC15041-1000 AC15041-0250
200010-264EA 49.3 USD
200010-264 200005-396
α,4-Dibromotoluene 98%
α,4-Dibromotoluene
Formula: C₇H₆Br₂
MW: 249.93 g/mol
Boiling Pt: 115…124 °C (12 mmHg)
Melting Pt: 60…64 °C
Storage Temperature: Ambient
MDL Number: MFCD00000179
CAS Number: 589-15-1
EINECS: 209-636-5
UN: 1759
ADR: 8,II
Merck Index: 13,01393

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR