(±)-2-Methyl-2,4-pentanediol 99%

Warning

Synonyms: (±)-2,4-Dihydroxy-2-methyl pentane2-Methylpentane-2,4-diolHexylene GlycolHexylene glycol

AC15034-0025 AC15034-0010 AC15034-0250
103464-768EA 82.6 USD
103464-768 103466-782 200001-158
(±)-2-Methyl-2,4-pentanediol 99%
(±)-2-Methyl-2,4-pentanediol
Formula: C₆H₁₄O₂
MW: 118.18 g/mol
Boiling Pt: 197 °C (760 mmHg)
Melting Pt: –40 °C
Density: 0.922 g/cm³
Flash Pt: 93 °C
Storage Temperature: Ambient
MDL Number: MFCD00004547
CAS Number: 107-41-5
EINECS: 203-489-0
Merck Index: 13,04730

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR