2-Methyl-2-propen-1-ol 98%

Warning

Synonyms: 2-Methyl allyl alcohol3-Hydroxy-2-methylpropene2-Methylprop-2-en-1-ol2-Methyl-2-propen-1-ol

AC15029-0100 AC15029-1000 AC15029-5000
200005-104EA 65.1 USD
200005-104 103525-760 103525-762
2-Methyl-2-propen-1-ol 98%
2-Methyl-2-propen-1-ol
Formula: C₄H₈O
MW: 72.11 g/mol
Boiling Pt: 113…115 °C (760 mmHg)
Melting Pt: –50 °C
Density: 0.85 g/cm³
Flash Pt: 33 °C
Storage Temperature: Ambient
MDL Number: MFCD00004737
CAS Number: 513-42-8
EINECS: 208-161-0
UN: 2614
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR