Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

α,4-Dichlorotoluene ≥99%

Supplier: ACROS ORGANICS MS

Warning
Synonyms: 4-Chlorobenzyl chloride1-Chloro-4-(chloromethyl)benzene

AC15024-0050 AC15024-2500 AC15024-0010
200010-088EA 25.42 USD InStock
200010-088 200010-348 103531-326
α,4-Dichlorotoluene ≥99%
α,4-Dichlorotoluene


Formula: C₇H₆Cl₂
MW: 161.03 g/mol
Boiling Pt: 221 °C (760 mmHg)
Melting Pt: 27…28 °C
Density: 1.259 g/cm³ (25 °C)
Flash Pt: 107 °C
Storage Temperature: Ambient
MDL Number: MFCD00000914
CAS Number: 104-83-6
EINECS: 203-242-7
UN: 3427
ADR: 6.1,III
Order Now
Specification Test Results
Appearance Colorless to white Low melting crystalline solid
Infrared spectrum Conforms
Melting point ≥28°C
GC ≥99.0 %
GC Impurities profile
GC ≤0.5 % p-Chlorotoluene
GC ≤0.5 % p-Chlorobenzal chloride


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty