3,4-Dichlorobenzoyl chloride 97%

Danger

AC15023-0250 AC15023-1000 AC15023-0050
200023-026EA 50.4 USD
200023-026 200029-052 200023-028
3,4-Dichlorobenzoyl chloride 97%
3,4-Dichlorobenzoyl chloride
Formula: C₇H₃Cl₃O
MW: 209.46 g/mol
Boiling Pt: 242 °C (1013 hPa)
Melting Pt: 30…33 °C
Flash Pt: 142 °C
Storage Temperature: Ambient
MDL Number: MFCD00000672
CAS Number: 3024-72-4
EINECS: 221-177-2
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR