4-Chlorobenzoyl chloride 99+%

Danger

Synonyms: p-Chlorobenzoyl chloride

AC15019-5000 AC15019-0050 AC15019-1000 AC15019-0025
200005-422EA 109 USD
200005-422 200010-344 200010-342 103525-758
4-Chlorobenzoyl chloride 99+%
4-Chlorobenzoyl chloride
Formula: C₇H₄Cl₂O
MW: 175.01 g/mol
Boiling Pt: 222 °C (760 mmHg)
Melting Pt: 12…14 °C
Density: 1.37 g/cm³
Flash Pt: 105 °C
Storage Temperature: Ambient
MDL Number: MFCD00000686
CAS Number: 122-01-0
EINECS: 204-515-3
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR